Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COBRA1 Mouse Monoclonal Antibody [clone: F7E4]

Supplier: Rockland Immunochemicals
200-301-D18
RL200-301-D18EA 528 USD InStock
RL200-301-D18
Anti-COBRA1 Mouse Monoclonal Antibody [clone: F7E4]
Antibodies
Anti-COBRA 1 antibody is suitable for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 62 kDa in size corresponding to COBRA1 Proteins in neonatal Rat brain lysate.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000

Type: Primary
Antigen: COBRA1
Clonality: Monoclonal
Clone: F7E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty