Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIA1 Mouse Monoclonal Antibody [clone: RH95]

Supplier: Rockland Immunochemicals
200-301-D61
RL200-301-D61EA 528 USD InStock
RL200-301-D61
Anti-GRIA1 Mouse Monoclonal Antibody [clone: RH95]
Antibodies
Anti-GluR1 (Mouse) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 105 kDa in size corresponding to GluR1 Proteins in Western Blot.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GRIA1
Clonality: Monoclonal
Clone: RH95
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty