Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RXRB Mouse Monoclonal Antibody [clone: 147]

Supplier: Rockland Immunochemicals
200-301-E27
RL200-301-E27EA 528 USD InStock
RL200-301-E27
Anti-RXRB Mouse Monoclonal Antibody [clone: 147]
Antibodies
Anti-Retinoid X Receptor beta antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 48 kDa in size corresponding to the RXR-beta Protein in the appropriate cell lysate.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RXRB
Clonality: Monoclonal
Clone: 147
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty