Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STX1A Mouse Monoclonal Antibody [clone: SP8]

Supplier: Rockland Immunochemicals
200-301-E40
RL200-301-E40EA 528 USD InStock
RL200-301-E40
Anti-STX1A Mouse Monoclonal Antibody [clone: SP8]
Antibodies
Anti-Syntaxin antibody is suitable for use in Western Blotting and IHC. Expect a band or approximately 36 kDa in size corresponding to syntaxin Proteins in Western blot of Rat brain lysate.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100

Type: Primary
Antigen: STX1A
Clonality: Monoclonal
Clone: SP8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Pig, Hamster

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty