Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-YDJ1 Mouse Monoclonal Antibody [clone: 1G10.H8]

Supplier: Rockland Immunochemicals
200-301-E58
RL200-301-E58EA 532 USD InStock
RL200-301-E58
Anti-YDJ1 Mouse Monoclonal Antibody [clone: 1G10.H8]
Antibodies
Anti-Hsp40 Antibody is recommended for use in WB, IP, and ELISA. Expect a band approximately ~40kDa Protein corresponding to the molecular mass of Hsp40 on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000;

Type: Primary
Antigen: YDJ1
Clonality: Monoclonal
Clone: 1G10.H8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Yeast

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty