Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRYAA Mouse Monoclonal Antibody [clone: 1H3.B8]

Supplier: Rockland Immunochemicals
200-301-E84
RL200-301-E84EA 532 USD InStock
RL200-301-E84
Anti-CRYAA Mouse Monoclonal Antibody [clone: 1H3.B8]
Antibodies
Anti-Alpha A Crystallin Antibody is recommended for use in WB and ELISA. Expect a band approximately ~20kDa Protein corresponding to the molecular mass of a-crystallin on SDS PAGE immunoblots.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000

Type: Primary
Antigen: CRYAA
Clonality: Monoclonal
Clone: 1H3.B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty