Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRYAB Mouse Monoclonal Antibody [clone: 3A10.H4]

Supplier: Rockland Immunochemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

200-301-E86 200-301-E85
RL200-301-E86EA 532 USD InStock
RL200-301-E86 RL200-301-E85
Anti-CRYAB Mouse Monoclonal Antibody [clone: 3A10.H4]
Antibodies
Anti-Alpha B-crystallin Antibody is recommended for use in WB and ELISA. Expect a band approximately ~20kDa (predicted mol.weight is ~21kDa) corresponding to aB-crystallin. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 3A10.H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty