Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KCNMB4 Mouse Monoclonal Antibody [clone: S18A-3]

Supplier: Rockland Immunochemicals
200-301-E94
RL200-301-E94EA 532 USD InStock
RL200-301-E94
Anti-KCNMB4 Mouse Monoclonal Antibody [clone: S18A-3]
Antibodies
Anti-BK Beta4 Antibody is suitable for use in WB, IP, and IHC. Antibody is provided in PBS pH 7.4. Expect a band approximately ~24kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions:

Type: Primary
Antigen: KCNMB4
Clonality: Monoclonal
Clone: S18A-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty