Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CACNA1C Mouse Monoclonal Antibody [clone: S57-46]

Supplier: Rockland Immunochemicals
200-301-F08
RL200-301-F08EA 532 USD InStock
RL200-301-F08
Anti-CACNA1C Mouse Monoclonal Antibody [clone: S57-46]
Antibodies
Anti-CAV1.2 Antibody is suitable for use in WB, IP and IHC. Expect a band approximately ~240kDa (varies with cell background due to glycosylation) on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions:

Type: Primary
Antigen: CACNA1C
Clonality: Monoclonal
Clone: S57-46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty