Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DMPO Mouse Monoclonal Antibody [clone: N1664A]

Supplier: Rockland Immunochemicals
200-301-F21
RL200-301-F21EA 532 USD InStock
RL200-301-F21
Anti-DMPO Mouse Monoclonal Antibody [clone: N1664A]
Antibodies
Anti-DMPO Antibody is suitable for use in Immuno-spin trapping, ELISA, WB, Immunofluorescence microscopy and IP. Expect a band approximately ~90kDa corresponding to specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1-10µg/ml; Immunofluorescence: 10µg/ml; Immunoprecipitation: 25µg

Type: Primary
Antigen: DMPO
Clonality: Monoclonal
Clone: N1664A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pan, Adenovirus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty