Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOD3 Mouse Monoclonal Antibody [clone: 4GG11G6]

Supplier: Rockland Immunochemicals
200-301-F24
RL200-301-F24EA 532 USD InStock
RL200-301-F24
Anti-SOD3 Mouse Monoclonal Antibody [clone: 4GG11G6]
Antibodies
Anti-EC SOD Antibody is recommended for use in WB, Immunochemistryand ELISA. Expect a band approximately ~35kDa in specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Immunofluorescence: 1:100; Western Blot: 1:1000; Immunochemistry: 1:100

Type: Primary
Antigen: SOD3
Clonality: Monoclonal
Clone: 4GG11G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty