Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPA5 Mouse Monoclonal Antibody [clone: 1H11-1H7]

Supplier: Rockland Immunochemicals
200-301-F36
RL200-301-F36EA 532 USD InStock
RL200-301-F36
Anti-HSPA5 Mouse Monoclonal Antibody [clone: 1H11-1H7]
Antibodies
Type: Primary
Antigen: HSPA5
Clonality: Monoclonal
Clone: 1H11-1H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Bovine, Cryphonectria Parasitica, Hamster, Monkey, Rabbit, Xenopus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty