Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPB8 Mouse Monoclonal Antibody [clone: 3C12-H11]

Supplier: Rockland Immunochemicals
200-301-F53
RL200-301-F53EA 532 USD InStock
RL200-301-F53
Anti-HSPB8 Mouse Monoclonal Antibody [clone: 3C12-H11]
Antibodies
Anti-Hsp22 Antibody is suitable for use inWB, Immunochemistryand ELISA. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000;

Type: Primary
Antigen: HSPB8
Clonality: Monoclonal
Clone: 3C12-H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty