Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPB2 Mouse Monoclonal Antibody [clone: 8A7]

Supplier: Rockland Immunochemicals
200-301-F55
RL200-301-F55EA 532 USD InStock
RL200-301-F55
Anti-HSPB2 Mouse Monoclonal Antibody [clone: 8A7]
Antibodies
Anti-Hsp25/Hsp27 Antibody is suitable for WB, IF, Immunofluorescence microscopy, IP, and IHC. Expect a band approximately 25kDa or 27kDa Protein corresponding to the molecular mass of Hsp25/27 on SDS PAGE immunoblots.

Recommended Dilutions:

Type: Primary
Antigen: HSPB2
Clonality: Monoclonal
Clone: 8A7
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Rat, Bovine, Dog, Guinea Pig, Hamster

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty