Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPD1 Mouse Monoclonal Antibody [clone: LK2]

Supplier: Rockland Immunochemicals
200-301-F61
RL200-301-F61EA 532 USD InStock
RL200-301-F61
Anti-HSPD1 Mouse Monoclonal Antibody [clone: LK2]
Antibodies
Anti-Hsp60 Antibody is suitable for WB, Flow Cytometry, and IHC. Expect a band approximately ~60kDa Protein corresponding to the molecular mass of Hsp60 on SDS PAGE immunoblots.

Recommended Dilutions: Western Blot: 0.25 µg/ml

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: LK2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken, Guinea Pig, Hamster, Monkey, Pig, Rabbit, Spinach, E. coli, H. Pylori, S. Typhimurium, T. Spiralis , Yeast, White Fly

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty