Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPA1A Mouse Monoclonal Antibody [clone: BB70]

Supplier: Rockland Immunochemicals
200-301-F64
RL200-301-F64EA 537 USD InStock
RL200-301-F64
Anti-HSPA1A Mouse Monoclonal Antibody [clone: BB70]
Antibodies
Anti-Hsp70/Hsc70 Antibody is suitable for WB, IP, and IHC. Expect a band approximately ~72 and ~73kDa Proteins corresponding to the molecular mass of inducible Hsp and Hsc on SDS PAGE immunoblots.

Recommended Dilutions: Western Blot: 1µg/ml;

Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: BB70
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Sheep, Dog, Beluga, Bovine, Fish, Guinea Pig, Scallop Pig, Hamster, Rabbit, Chicken, Xenopus, Drosophila, Yeast

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty