Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP70 Mouse Monoclonal Antibody [clone: 5G1-95]

Supplier: Rockland Immunochemicals
200-301-F65
RL200-301-F65EA 532 USD InStock
RL200-301-F65
Anti-HSP70 Mouse Monoclonal Antibody [clone: 5G1-95]
Antibodies
Anti-Hsp70/Hsc70 Antibody is suitable for WB and ELISA. Expect a band approximately ~70kDa corresponding to the molecular mass Hsp70 on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200

Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone: 5G1-95
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Plants, Hantavirus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty