Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 8D3]

Supplier: Rockland Immunochemicals
200-301-F73
RL200-301-F73EA 671 USD InStock
RL200-301-F73
Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 8D3]
Antibodies
Anti-Hsp90 complex Antibody is suitable for IP. Expect a band approximately 90kDa Proteins corresponding to the molecular mass of Hsp90 for immunoprecitpitation.

Recommended Dilutions:

Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: 8D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty