Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAMP1 Mouse Monoclonal Antibody [clone: Ly1C6]

Supplier: Rockland Immunochemicals
200-301-G09
RL200-301-G09EA 532 USD InStock
RL200-301-G09
Anti-LAMP1 Mouse Monoclonal Antibody [clone: Ly1C6]
Antibodies
Anti-LAMP1 Antibody is suitable for WB, IP, and Immunofluorescence microscopy. Expect a band approximately ~120kDa Protein corresponding to the molecular mass of LAMP1 on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1µg/ml;

Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: Ly1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty