Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SCN2A Mouse Monoclonal Antibody [clone: S69-3]

Supplier: Rockland Immunochemicals
200-301-G19
RL200-301-G19EA 532 USD InStock
RL200-301-G19
Anti-SCN2A Mouse Monoclonal Antibody [clone: S69-3]
Antibodies
Anti-Nav1.2 Antibody is suitable for use in WB, IP, and IHC. Expect a band approximately ~250kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions:

Type: Primary
Antigen: SCN2A
Clonality: Monoclonal
Clone: S69-3
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty