Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTGES3 Mouse Monoclonal Antibody [clone: JJ6]

Supplier: Rockland Immunochemicals
200-301-G25
RL200-301-G25EA 532 USD InStock
RL200-301-G25
Anti-PTGES3 Mouse Monoclonal Antibody [clone: JJ6]
Antibodies
Anti-p23 Antibody is recommended for use in WB, IP, and ELISA. Expect a band approximately ~23kDa Protein corresponding to the molecular mass of p23 on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000

Type: Primary
Antigen: PTGES3
Clonality: Monoclonal
Clone: JJ6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouses, Rabbit, Chicken, Guinea Pig, S. Cerevisiae (Lower)

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty