Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OLFM1 Mouse Monoclonal Antibody [clone: S96-7]

Supplier: Rockland Immunochemicals
200-301-G28
RL200-301-G28EA 532 USD InStock
RL200-301-G28
Anti-OLFM1 Mouse Monoclonal Antibody [clone: S96-7]
Antibodies
Anti-Pancortin Antibody is suitable for use in WB, Immunochemistryand IP. Expect a band approximately ~70kDa (Pancortin-1, Pancortin-3) and ~20kDa (Pancortin-2 and Pancortin-4) on specific lysates.

Recommended Dilutions:

Type: Primary
Antigen: OLFM1
Clonality: Monoclonal
Clone: S96-7
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty