Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DLG4 Mouse Monoclonal Antibody [clone: 6G6]

Supplier: Rockland Immunochemicals
200-301-G32
RL200-301-G32EA 532 USD InStock
RL200-301-G32
Anti-DLG4 Mouse Monoclonal Antibody [clone: 6G6]
Antibodies
Type: Primary
Antigen: DLG4
Clonality: Monoclonal
Clone: 6G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Bovine, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty