Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RHO Mouse Monoclonal Antibody [clone: 1D4]

Supplier: Rockland Immunochemicals
200-301-G37
RL200-301-G37EA 536 USD InStock
RL200-301-G37
Anti-RHO Mouse Monoclonal Antibody [clone: 1D4]
Antibodies
Anti-Rhodopsin Antibody is recommended for use in Immunofluorescence microscopy, IHC, IP, WB, and ELISA. Expect a band approximately ~40kDa corresponding to specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:1000;

Type: Primary
Antigen: RHO
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Vertebrates, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty