Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UBB Mouse Monoclonal Antibody [clone: 5B9-B3]

Supplier: Rockland Immunochemicals
200-301-G62 200-301-G61
RL200-301-G62EA 532 USD InStock
RL200-301-G62 RL200-301-G61
Anti-UBB Mouse Monoclonal Antibody [clone: 5B9-B3]
Antibodies
Anti-Ubiquitin Antibody is recommended for use in WB and ELISA. Expect a band approximately ~10kDa corresponding to free ubiquitin. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:1000

Type: Primary
Antigen: UBB
Clonality: Monoclonal
Clone: 5B9-B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty