Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GATA4 Mouse Monoclonal Antibody [clone: 10D8.B10.D4]

Supplier: Rockland Immunochemicals
200-301-P15 200-301-P15
RL200-301-P15EA 528 USD InStock
RL200-301-P15 CA200-301-P15
Anti-GATA4 Mouse Monoclonal Antibody [clone: 10D8.B10.D4]
Antibodies
Anti-GATA4 antibody is suitable for ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 44 kDa in size corresponding to GATA4 by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: GATA4
Clonality: Monoclonal
Clone: 10D8.B10.D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty