Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD47 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 15F6.F6.F9]

Supplier: Rockland Immunochemicals
200-302-M68 200-302-M68
RL200-302-M68EA 693 USD InStock
RL200-302-M68 CA10036-884
Anti-CD47 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 15F6.F6.F9]
Antibodies
Anti-CD47 is useful for Flow Cytometry (Cell Surface). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/5 x 10^5 cells (0.5 µg)
Do not freeze!

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: 15F6.F6.F9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty