Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL2RA Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 9G4.C3.F2]

Supplier: Rockland Immunochemicals
200-302-N25 200-302-N25
RL200-302-N25EA 285 USD InStock
CA10036-896 RL200-302-N25
Anti-IL2RA Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 9G4.C3.F2]
Antibodies
Anti-CD25 is useful for Flow Cytometry and Immunofluorescence. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/1x10e6 cells or 100µl of whole blood;
Do not freeze!

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 9G4.C3.F2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty