Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD27 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 11D9.F3.C12.B8]

Supplier: Rockland Immunochemicals
200-302-N65 200-302-N65
RL200-302-N65EA 440 USD InStock
CA10036-906 RL200-302-N65
Anti-CD27 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 11D9.F3.C12.B8]
Antibodies
Anti-CD27 is useful for Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/1x10e6 cells or 100µl of whole blood
Do not freeze!

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 11D9.F3.C12.B8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty