Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 5C3]

Supplier: Rockland Immunochemicals
200-302-N67 200-302-N67
RL200-302-N67EA 440 USD InStock
CA200-302-N67 RL200-302-N67
Anti-CD40 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 5C3]
Antibodies
Anti-CD40 is useful for Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells
Do not freeze!

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 5C3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty