Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATM Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10H11.E12]

Supplier: Rockland Immunochemicals
200-303-400 200-303-400
RL200-303-400EA 615 USD InStock
RL200-303-400 CA200-303-400
Anti-ATM Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10H11.E12]
Antibodies
This product has been tested by ELISA and western blotting against both the native and recombinant forms of the Protein. This reagent may also be suitable for immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibodies.

Recommended Dilutions: ELISA: 1:70,000; Western Blot: 1:600;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone: 10H11.E12
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty