Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-6xHis Mouse Monoclonal Antibody (Biotin) [clone: 14A231]

Supplier: Rockland Immunochemicals
200-306-382
RL200-306-382EA 502 USD InStock
RL200-306-382
Anti-6xHis Mouse Monoclonal Antibody (Biotin) [clone: 14A231]
Antibodies
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: 14A231
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty