Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD8A Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: OKT8]

Supplier: Rockland Immunochemicals
200-308-N62 200-308-N62
RL200-308-N62EA 275 USD InStock
RL200-308-N62 CA10036-938
Anti-CD8A Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: OKT8]
Antibodies
Anti-CD8a is useful for immunochemistry and Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5µl/test/1x10e5 to 1x10e8 cells;
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: OKT8
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty