Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTPRC Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HI30]

Supplier: Rockland Immunochemicals
200-308-N68 200-308-N68
RL200-308-N68EA 364 USD InStock
CA200-308-N68 RL200-308-N68
Anti-PTPRC Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HI30]
Antibodies
Anti-CD45 is useful for Flow Cytometry, immunochemistry, and Western Blotting. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze! Light sensitive!

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI30
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty