Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 14E5.A2.B2.H9]

Supplier: Rockland Immunochemicals
200-326-I51 200-326-I51
RL200-326-I51EA 539 USD InStock
RL200-326-I51 CA200-326-I51
Anti-AKT1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 14E5.A2.B2.H9]
Antibodies
Anti-AKT1 APC Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT1 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: Flow Cytometry: 1.0 µg/ml;
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.A2.B2.H9
Conjugation: APC (Allophycocyanin)
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty