Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT3a]

Supplier: Rockland Immunochemicals
200-326-N12 200-326-N12
RL200-326-N12EA 399 USD InStock
CA10036-956 RL200-326-N12
Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT3a]
Antibodies
Anti-CD3 is useful for Immunoprecipitation, immunochemistry, and Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/1x10e6 cells or 100µl of whole blood;
Do not freeze!

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: HIT3a
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty