Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SELL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: DREG-56]

Supplier: Rockland Immunochemicals
200-326-N72 200-326-N72
RL200-326-N72EA 632 USD InStock
RL200-326-N72 CA200-326-N72
Anti-SELL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: DREG-56]
Antibodies
Anti-CD62L is useful for Flow Cytometry and Western Blot. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions:
Do not freeze!

Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: DREG-56
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty