Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: IT2.2]

Supplier: Rockland Immunochemicals
200-326-N86 200-326-N86
RL200-326-N86EA 539 USD InStock
CA200-326-N86 RL200-326-N86
Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: IT2.2]
Antibodies
Anti-CD45.2 is useful for immunochemistry, Immunoprecipitation, Immunofluorescence, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: IT2.2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty