Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 649) [clone: 18F3.H11]

Supplier: Rockland Immunochemicals
200-343-269
RL200-343-269EA 508 USD InStock
RL200-343-269
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 649) [clone: 18F3.H11]
Antibodies
This monoclonal antibody is suitable for immunofluorescence microscopy, FLISA and western blotting. Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:4000-1:20000; FLISA: >1:20000; Western Blot: 1:10000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 18F3.H11
Conjugation: DyLight® 649
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty