Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OVAL Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-033 200-401-033S
RL200-401-033EA 523 USD InStock
RL200-401-033 RL200-401-033S
Anti-OVAL Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:300 - 12000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: OVAL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty