Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AMY Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-045 200-401-045S
RL200-401-045EA 518 USD InStock
RL200-401-045 RL200-401-045S
Anti-AMY Rabbit Polyclonal Antibody
Antibodies
This product has been assayed against 1.0 µg of a-Amylase (Bacillus amyloliquefaciens) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: AMY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacillus amyloliquefaciens

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty