Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CIITA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-192 200-401-192
RL200-401-192EA 518 USD InStock
CA200-401-192 RL200-401-192
Anti-CIITA Rabbit Polyclonal Antibody
Antibodies
Anti-CIITA is suitable for the detection by immunoblot of human CIITA and ELISA.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: CIITA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty