Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP3K5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-410 200-401-410
RL200-401-410EA 518 USD InStock
CA200-401-410 RL200-401-410
Anti-MAP3K5 Rabbit Polyclonal Antibody
Antibodies
This phospho specific polyclonal antibody reacts human pS83 ASK1 and shows minimal reactivity by western blot, ELISA and competitive ELISA with non-phosphorylated ASK1. Although not tested, this antibody is likely functional in RIA.

Recommended Dilutions: ELISA: 1:5000 - 1:10000; Western Blot: 1:1000;

Type: Primary
Antigen: MAP3K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty