Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RUB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-427 200-401-427
RL200-401-427EA 570 USD InStock
CA200-401-427 RL200-401-427
Anti-RUB1 Rabbit Polyclonal Antibody
Antibodies
Rub1 Antibody is deected as a 6 kDa band corresponding to yeast Rub1in immunoblotting.  Most yeast cell lysates can be used as a positive control without induction or stimulation.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 500 µl

Type: Primary
Antigen: RUB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty