Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UBB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-431 200-401-431
RL200-401-431EA 575 USD InStock
CA200-401-431 RL200-401-431
Anti-UBB Rabbit Polyclonal Antibody
Antibodies
This purified polyclonal antibody reacts with ubiquitin by ELISA and western blot. Although not tested, this antibody is likely functional in immunochemistry and immunoprecipitation. For detection of free Ub by western blotting use Tris-Tricine SDS-PAGE.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:200 - 1:1000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: UBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Many Species

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty