Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ISG15 Rabbit Monoclonal Antibody [clone: 1H12]

Supplier: Rockland Immunochemicals
200-401-438 200-401-438
RL200-401-438EA 570 USD InStock
CA200-401-438 RL200-401-438
Anti-ISG15 Rabbit Monoclonal Antibody [clone: 1H12]
Antibodies
This purified polyclonal antibody reacts with human ISG15 by western blot and ELISA. Although not tested, this antibody is likely functional in immunochemistry and immunoprecipitation.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:200 - 1:1000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: ISG15
Clonality: Monoclonal
Clone: 1H12
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty