Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SUMO1 Rabbit Polyclonal Antibody [clone: G3.1]

Supplier: Rockland Immunochemicals
200-401-441 200-401-441
RL200-401-441EA 570 USD InStock
CA200-401-441 RL200-401-441
Anti-SUMO1 Rabbit Polyclonal Antibody [clone: G3.1]
Antibodies
This purified polyclonal antibody reacts with human SUMO by western blot and ELISA.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: SUMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty