Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NRF1 Rabbit Polyclonal Antibody [clone: 7F8]

Supplier: Rockland Immunochemicals
200-401-869 200-401-869
RL200-401-869EA 570 USD InStock
CA200-401-869 RL200-401-869
Anti-NRF1 Rabbit Polyclonal Antibody [clone: 7F8]
Antibodies
Type: Primary
Antigen: NRF1
Clonality: Polyclonal
Clone: 7F8
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty