Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCNL1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-881 200-401-881
RL200-401-881EA 570 USD InStock
CA200-401-881 RL200-401-881
Anti-CCNL1 Rabbit Polyclonal Antibody
Antibodies
This Protein A purified antibody has been tested for use in ELISA, immunochemistry western blot, immunoprecipitation and immunofluores-cence.   Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 59 kDa in size.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:2000 - 1:5000; Immunochemistry: 10 mg/ml - 20 µg/ml; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: CCNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty