Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADGRE5 Rabbit Polyclonal Antibody [clone: 1H11.E12]

Supplier: Rockland Immunochemicals
200-401-908 200-401-908
RL200-401-908EA 570 USD InStock
CA200-401-908 RL200-401-908
Anti-ADGRE5 Rabbit Polyclonal Antibody [clone: 1H11.E12]
Antibodies
This Protein A purified antibody has been tested for use in immunofluorescence microscopy and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 81 kDa in size.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Western Blot: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: ADGRE5
Clonality: Polyclonal
Clone: 1H11.E12
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty